Assay Details
| Gene Symbol: cxxc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200333.1 | NP_956627.1 | 11 - 12 | 1462 | 68 | |
| GenBank mRNA | BC052918.1 | - | 11 - 12 | 1462 | 68 | |
| BC165319.1 | - | 10 - 11 | 1327 | 68 | ||
More Information
Set Membership: |
|

cxxc1b
Genomic Map