Assay Details
| Gene Symbol: cxxc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200333.1 | NP_956627.1 | 12 - 13 | 1560 | 122 | |
| GenBank mRNA | BC052918.1 | - | 12 - 13 | 1560 | 122 | |
| BC165319.1 | - | 11 - 12 | 1425 | 122 | ||
More Information
Set Membership: |
|

cxxc1b
Genomic Map