Assay Details
| Gene Symbol: cxxc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200333.1 | NP_956627.1 | 13 - 14 | 1719 | 96 | |
| GenBank mRNA | BC052918.1 | - | 13 - 14 | 1719 | 96 | |
| BC165319.1 | - | 12 - 13 | 1584 | 96 | ||
More Information
Set Membership: |
|

cxxc1b
Genomic Map