Assay Details
| Gene Symbol: maco1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200463.2 | NP_956757.2 | 4 - 5 | 593 | 122 | |
| XM_005157048.4 | XP_005157105.1 | 4 - 5 | 572 | 122 | ||
| XM_021480543.1 | XP_021336218.1 | 4 - 5 | 680 | 122 | ||
| GenBank mRNA | AY845030.1 | - | 4 - 5 | 593 | 122 | |
More Information
Set Membership: |
|

maco1b
Genomic Map