Assay Details
| Gene Symbol: maco1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200463.2 | NP_956757.2 | 8 - 9 | 1720 | 104 | |
| XM_005157048.4 | XP_005157105.1 | 8 - 9 | 1696 | 104 | ||
| XM_021480543.1 | XP_021336218.1 | 8 - 9 | 1801 | 104 | ||
| GenBank mRNA | AY845030.1 | - | 8 - 9 | 1720 | 104 | |
| BC171392.1 | - | 8 - 9 | 1690 | 104 | ||
More Information
Set Membership: |
|

maco1b
Genomic Map