Assay Details
| Gene Symbol: slc25a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200607.1 | NP_956901.1 | 3 - 4 | 573 | 86 | |
| GenBank mRNA | BC056787.1 | - | 3 - 4 | 573 | 86 | |
More Information
Set Membership: |
|

slc25a1a
Genomic Map