Assay Details
| Gene Symbol: aanat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200704.1 | NP_956998.1 | 1 - 2 | 229 | 68 | |
| GenBank mRNA | AY349158.1 | - | 1 - 2 | 244 | 68 | |
| BC059448.1 | - | 1 - 2 | 229 | 68 | ||
More Information
Set Membership: |
|

aanat1
Genomic Map