Assay Details
| Gene Symbol: aanat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200704.1 | NP_956998.1 | 2 - 3 | 387 | 114 | |
| GenBank mRNA | AY349158.1 | - | 2 - 3 | 402 | 114 | |
| BC059448.1 | - | 2 - 3 | 387 | 114 | ||
More Information
Set Membership: |
|

aanat1
Genomic Map