Assay Details
| Gene Symbol: slc25a3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328572.1 | NP_001315501.1 | 3 - 4 | 354 | 100 | |
| NM_200715.2 | NP_957009.1 | 3 - 4 | 318 | 100 | ||
| XM_005164728.4 | XP_005164785.1 | 3 - 4 | 520 | 100 | ||
| GenBank mRNA | BC059476.1 | - | 3 - 4 | 295 | 100 | |
| EH451459.1 | - | 3 - 4 | 344 | 100 | ||
More Information
Set Membership: |
|

slc25a3a
Genomic Map