Assay Details
| Gene Symbol: slc25a3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328572.1 | NP_001315501.1 | 7 - 8 | 1004 | 80 | |
| NM_200715.2 | NP_957009.1 | 7 - 8 | 968 | 80 | ||
| XM_005164728.4 | XP_005164785.1 | 7 - 8 | 1170 | 80 | ||
| GenBank mRNA | BC059476.1 | - | 7 - 8 | 945 | 80 | |
| EH478503.1 | - | 1 - 2 | 417 | 80 | ||
| EH609753.1 | - | 1 - 2 | 417 | 80 | ||
More Information
Set Membership: |
|

slc25a3a
Genomic Map