Assay Details
| Gene Symbol: calb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200718.1 | NP_957012.1 | 1 - 2 | 144 | 80 | |
| GenBank mRNA | BC059479.1 | - | 1 - 2 | 144 | 80 | |
More Information
Set Membership: |
|

calb2a
Genomic Map