Assay Details
| Gene Symbol: arl3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200719.1 | NP_957013.1 | 3 - 4 | 386 | 80 | |
| XM_005169564.4 | XP_005169621.1 | 3 - 4 | 450 | 80 | ||
| GenBank mRNA | BC059480.1 | - | 3 - 4 | 386 | 80 | |
More Information
Set Membership: |
|

arl3l2
Genomic Map