Assay Details
| Gene Symbol: arl3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200719.1 | NP_957013.1 | 4 - 5 | 439 | 70 | |
| GenBank mRNA | BC059480.1 | - | 4 - 5 | 439 | 70 | |
More Information
Set Membership: |
|

arl3l2
Genomic Map