Assay Details
| Gene Symbol: fgf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200760.1 | NP_957054.1 | 2 - 3 | 333 | 133 | |
| GenBank mRNA | AB194698.1 | - | 2 - 3 | 237 | 133 | |
| BC059588.1 | - | 2 - 3 | 333 | 133 | ||
More Information
Set Membership: |
|

fgf1a
Genomic Map