Assay Details
| Gene Symbol: tfdp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200776.2 | NP_957070.2 | 3 - 4 | 278 | 107 | |
| XM_005167988.4 | XP_005168045.1 | 3 - 4 | 297 | 107 | ||
| XM_009305148.3 | XP_009303423.1 | 3 - 4 | 330 | 107 | ||
| GenBank mRNA | BC059610.1 | - | 3 - 4 | 237 | 107 | |
| EH541383.1 | - | 3 - 4 | 291 | 107 | ||
| EH593111.1 | - | 3 - 4 | 366 | 107 | ||
More Information
Set Membership: |
|

tfdp1b
Genomic Map