Assay Details
| Gene Symbol: eif3ea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200839.1 | NP_957133.1 | 8 - 9 | 868 | 87 | |
| GenBank mRNA | AY398340.1 | - | 8 - 9 | 859 | 87 | |
| BC061454.1 | - | 8 - 9 | 868 | 87 | ||
More Information
Set Membership: |
|

eif3ea
Genomic Map