Assay Details
| Gene Symbol: eif3ea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200839.1 | NP_957133.1 | 10 - 11 | 1084 | 90 | |
| GenBank mRNA | AY398340.1 | - | 10 - 11 | 1075 | 90 | |
| AY648778.1 | - | 10 - 11 | 1083 | 90 | ||
| BC061454.1 | - | 10 - 11 | 1084 | 90 | ||
More Information
Set Membership: |
|

eif3ea
Genomic Map