Assay Details
| Gene Symbol: srsf5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200867.1 | NP_957161.1 | 7 - 8 | 605 | 66 | |
| GenBank mRNA | AY398417.1 | - | 7 - 8 | 605 | 66 | |
| BC063235.1 | - | 6 - 7 | 66 | |||
More Information
Set Membership: |
|

srsf5a
Genomic Map