Assay Details
| Gene Symbol: arhgap12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200874.1 | NP_957168.1 | 13 - 14 | 2223 | 65 | |
| XM_009306595.3 | XP_009304870.1 | 14 - 15 | 2332 | 65 | ||
| XM_021480040.1 | XP_021335715.1 | 15 - 16 | 2455 | 65 | ||
| XM_021480041.1 | XP_021335716.1 | 15 - 16 | 2452 | 65 | ||
| XM_021480042.1 | XP_021335717.1 | 14 - 15 | 2383 | 65 | ||
| XM_021480043.1 | XP_021335718.1 | 14 - 15 | 1775 | 65 | ||
| GenBank mRNA | BC063321.1 | - | 13 - 14 | 2223 | 65 | |
| BC065619.1 | - | 14 - 15 | 3025 | 65 | ||
More Information
Set Membership: |
|

arhgap12b
Genomic Map