Assay Details
| Gene Symbol: arhgap12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200874.1 | NP_957168.1 | 16 - 17 | 2539 | 80 | |
| XM_009306595.3 | XP_009304870.1 | 17 - 18 | 2648 | 80 | ||
| XM_021480040.1 | XP_021335715.1 | 18 - 19 | 2771 | 80 | ||
| XM_021480041.1 | XP_021335716.1 | 18 - 19 | 2768 | 80 | ||
| XM_021480042.1 | XP_021335717.1 | 17 - 18 | 2699 | 80 | ||
| XM_021480043.1 | XP_021335718.1 | 17 - 18 | 2091 | 80 | ||
| GenBank mRNA | BC063321.1 | - | 16 - 17 | 2539 | 80 | |
| BC065619.1 | - | 17 - 18 | 3341 | 80 | ||
More Information
Set Membership: |
|

arhgap12b
Genomic Map