Assay Details
| Gene Symbol: ppp2cab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200911.3 | NP_957205.2 | 1 - 2 | 309 | 78 | |
| GenBank mRNA | BC045892.1 | - | 1 - 2 | 310 | 78 | |
| BC165877.1 | - | 1 - 2 | 130 | 78 | ||
More Information
Set Membership: |
|

ppp2cab
Genomic Map