Assay Details
| Gene Symbol: ppp2cab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200911.3 | NP_957205.2 | 2 - 3 | 526 | 105 | |
| GenBank mRNA | BC045892.1 | - | 2 - 3 | 527 | 105 | |
| BC165877.1 | - | 2 - 3 | 347 | 105 | ||
More Information
Set Membership: |
|

ppp2cab
Genomic Map