Assay Details
| Gene Symbol: sigmar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200977.1 | NP_957271.1 | 3 - 4 | 541 | 63 | |
| XM_009305185.1 | XP_009303460.1 | 2 - 3 | 149 | 63 | ||
| GenBank mRNA | BC049416.1 | - | 3 - 4 | 541 | 63 | |
More Information
Set Membership: |
|

sigmar1
Genomic Map