Assay Details
| Gene Symbol: srp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200988.2 | NP_957282.1 | 7 - 8 | 625 | 69 | |
| GenBank mRNA | BC045474.1 | - | 7 - 8 | 595 | 69 | |
| BC067588.1 | - | 7 - 8 | 610 | 69 | ||
| BC164217.1 | - | 5 - 6 | 465 | 69 | ||
| EH550891.1 | - | 7 - 8 | 647 | 69 | ||
More Information
Set Membership: |
|

srp54
Genomic Map