Assay Details
| Gene Symbol: rnf11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201021.1 | NP_957315.1 | 1 - 2 | 427 | 119 | |
| GenBank mRNA | BC053118.1 | - | 1 - 2 | 427 | 119 | |
More Information
Set Membership: |
|

rnf11a
Genomic Map