Assay Details
| Gene Symbol: prkcba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201029.1 | NP_957323.1 | 1 - 2 | 281 | 75 | |
| XM_005170844.4 | XP_005170901.1 | 1 - 2 | 326 | 75 | ||
| GenBank mRNA | BC054603.1 | - | 1 - 2 | 281 | 75 | |
| BC171680.1 | - | 1 - 2 | 245 | 75 | ||
More Information
Set Membership: |
|

prkcba
Genomic Map