Assay Details
| Gene Symbol: trim63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201095.1 | NP_957389.1 | 1 | 1050 | 72 | |
| GenBank mRNA | BC049414.1 | - | 1 | 1050 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

trim63b| Gene Symbol: trim63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201095.1 | NP_957389.1 | 1 | 1050 | 72 | |
| GenBank mRNA | BC049414.1 | - | 1 | 1050 | 72 | |
Set Membership: |
|