Assay Details
| Gene Symbol: acadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201181.2 | NP_957475.1 | 1 - 2 | 166 | 58 | |
| GenBank mRNA | BC044447.1 | - | 1 - 2 | 169 | 58 | |
| BC071366.1 | - | 1 - 2 | 193 | 58 | ||
| BC165576.1 | - | 1 - 2 | 125 | 58 | ||
More Information
Set Membership: |
|

acadl
Genomic Map