Assay Details
| Gene Symbol: rdh10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201331.1 | NP_958488.1 | 4 - 5 | 876 | 121 | |
| GenBank mRNA | AM422096.1 | - | 3 - 4 | 445 | 121 | |
| BC054596.1 | - | 4 - 5 | 876 | 121 | ||
| BC164370.1 | - | 4 - 5 | 786 | 121 | ||
More Information
Set Membership: |
|

rdh10b
Genomic Map