Assay Details
| Gene Symbol: rdh10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201331.1 | NP_958488.1 | 5 - 6 | 1013 | 142 | |
| GenBank mRNA | BC054596.1 | - | 5 - 6 | 1013 | 142 | |
| BC164370.1 | - | 5 - 6 | 923 | 142 | ||
More Information
Set Membership: |
|

rdh10b
Genomic Map