Assay Details
| Gene Symbol: pafah1b1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201346.1 | NP_958503.1 | 3 - 4 | 413 | 91 | |
| XM_021468786.1 | XP_021324461.1 | 3 - 4 | 337 | 91 | ||
| GenBank mRNA | BC044530.1 | - | 3 - 4 | 441 | 91 | |
| DQ141216.1 | - | 2 - 3 | 183 | 91 | ||
More Information
Set Membership: |
|

pafah1b1b
Genomic Map