Assay Details
| Gene Symbol: pafah1b1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201346.1 | NP_958503.1 | 4 - 5 | 500 | 115 | |
| XM_021468786.1 | XP_021324461.1 | 4 - 5 | 424 | 115 | ||
| GenBank mRNA | BC044530.1 | - | 4 - 5 | 528 | 115 | |
| DQ141216.1 | - | 3 - 4 | 270 | 115 | ||
More Information
Set Membership: |
|

pafah1b1b
Genomic Map