Assay Details
| Gene Symbol: pafah1b1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201346.1 | NP_958503.1 | 7 - 8 | 981 | 78 | |
| XM_021468786.1 | XP_021324461.1 | 7 - 8 | 78 | |||
| GenBank mRNA | BC044530.1 | - | 7 - 8 | 1009 | 78 | |
| DQ141216.1 | - | 6 - 7 | 78 | |||
More Information
Set Membership: |
|

pafah1b1b
Genomic Map