Assay Details
| Gene Symbol: ywhaqa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201513.1 | NP_958921.1 | 4 - 5 | 770 | 108 | |
| GenBank mRNA | BC048068.1 | - | 4 - 5 | 770 | 108 | |
| BC066409.1 | - | 3 - 4 | 745 | 108 | ||
| BC154307.1 | - | 4 - 5 | 766 | 108 | ||
More Information
Set Membership: |
|

ywhaqa
Genomic Map