Assay Details
| Gene Symbol: naa15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203321.2 | NP_976066.2 | 7 - 8 | 977 | 76 | |
| GenBank mRNA | AY391464.1 | - | 7 - 8 | 964 | 76 | |
| BC044392.1 | - | 7 - 8 | 894 | 76 | ||
| BC071438.1 | - | 7 - 8 | 977 | 76 | ||
More Information
Set Membership: |
|

naa15b
Genomic Map