Assay Details
| Gene Symbol: actn3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205544.2 | NP_991107.2 | 11 - 12 | 1442 | 94 | |
| XM_021477400.1 | XP_021333075.1 | 11 - 12 | 1445 | 94 | ||
| GenBank mRNA | AF524840.1 | - | 1433 | 94 | ||
| BC065595.1 | - | 11 - 12 | 1345 | 94 | ||
| FJ979802.1 | - | 11 - 12 | 1269 | 94 | ||
More Information
Set Membership: |
|

actn3b
Genomic Map