Assay Details
| Gene Symbol: actn3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205544.2 | NP_991107.2 | 20 - 21 | 2712 | 105 | |
| XM_021477400.1 | XP_021333075.1 | 20 - 21 | 2715 | 105 | ||
| GenBank mRNA | AF524840.1 | - | 2703 | 105 | ||
| BC065595.1 | - | 20 - 21 | 2615 | 105 | ||
| FJ979802.1 | - | 20 - 21 | 2539 | 105 | ||
More Information
Set Membership: |
|

actn3b
Genomic Map