Assay Details
| Gene Symbol: bnip3lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205571.2 | NP_991134.2 | 3 - 4 | 372 | 131 | |
| GenBank mRNA | AF493987.1 | - | 3 - 4 | 131 | ||
| BC091797.1 | - | 3 - 4 | 372 | 131 | ||
| BC165026.1 | - | 3 - 4 | 374 | 131 | ||
More Information
Set Membership: |
|

bnip3lb
Genomic Map