Assay Details
Gene Symbol: ankrd46b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205596.1 | NP_991159.1 | 2 - 3 | 511 | 68 | |
XM_005166503.3 | XP_005166560.1 | 3 - 4 | 614 | 68 | ||
GenBank mRNA | AY391435.1 | - | 2 - 3 | 511 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
