Assay Details
| Gene Symbol: u2af2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205689.1 | NP_991252.1 | 10 - 11 | 982 | 89 | |
| XM_009294031.2 | XP_009292306.1 | 11 - 12 | 1539 | 89 | ||
| XM_009294032.3 | XP_009292307.1 | 11 - 12 | 988 | 89 | ||
| XM_009294033.3 | XP_009292308.1 | 10 - 11 | 1386 | 89 | ||
| XM_009294034.3 | XP_009292309.1 | 10 - 11 | 1503 | 89 | ||
| XM_009294035.3 | XP_009292310.1 | 9 - 10 | 1488 | 89 | ||
| XM_009294036.3 | XP_009292311.1 | 9 - 10 | 1350 | 89 | ||
| XM_009294037.3 | XP_009292312.1 | 8 - 9 | 896 | 89 | ||
| GenBank mRNA | BC065869.1 | - | 10 - 11 | 982 | 89 | |
More Information
Set Membership: |
|

u2af2b
Genomic Map