Assay Details
| Gene Symbol: u2af2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205689.1 | NP_991252.1 | 12 - 13 | 1322 | 67 | |
| XM_009294031.2 | XP_009292306.1 | 13 - 14 | 1879 | 67 | ||
| XM_009294032.3 | XP_009292307.1 | 13 - 14 | 1328 | 67 | ||
| XM_009294033.3 | XP_009292308.1 | 12 - 13 | 1726 | 67 | ||
| XM_009294034.3 | XP_009292309.1 | 12 - 13 | 1843 | 67 | ||
| XM_009294035.3 | XP_009292310.1 | 11 - 12 | 1828 | 67 | ||
| XM_009294036.3 | XP_009292311.1 | 11 - 12 | 1690 | 67 | ||
| XM_009294037.3 | XP_009292312.1 | 10 - 11 | 1236 | 67 | ||
| GenBank mRNA | BC065869.1 | - | 12 - 13 | 1322 | 67 | |
More Information
Set Membership: |
|

u2af2b
Genomic Map