Assay Details
| Gene Symbol: u2af2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205689.1 | NP_991252.1 | 3 - 4 | 235 | 80 | |
| XM_009294031.2 | XP_009292306.1 | 3 - 4 | 777 | 80 | ||
| XM_009294032.3 | XP_009292307.1 | 3 - 4 | 229 | 80 | ||
| XM_009294033.3 | XP_009292308.1 | 2 - 3 | 624 | 80 | ||
| GenBank mRNA | BC065869.1 | - | 3 - 4 | 235 | 80 | |
More Information
Set Membership: |
|

u2af2b
Genomic Map