Assay Details
| Gene Symbol: u2af2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205689.1 | NP_991252.1 | 5 - 6 | 373 | 101 | |
| XM_009294031.2 | XP_009292306.1 | 5 - 6 | 915 | 101 | ||
| XM_009294032.3 | XP_009292307.1 | 5 - 6 | 367 | 101 | ||
| XM_009294033.3 | XP_009292308.1 | 4 - 5 | 762 | 101 | ||
| XM_009294034.3 | XP_009292309.1 | 4 - 5 | 879 | 101 | ||
| XM_009294035.3 | XP_009292310.1 | 4 - 5 | 879 | 101 | ||
| XM_009294036.3 | XP_009292311.1 | 3 - 4 | 726 | 101 | ||
| XM_009294037.3 | XP_009292312.1 | 3 - 4 | 287 | 101 | ||
| GenBank mRNA | BC065869.1 | - | 5 - 6 | 373 | 101 | |
More Information
Set Membership: |
|

u2af2b
Genomic Map