Assay Details
| Gene Symbol: u2af2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205689.1 | NP_991252.1 | 8 - 9 | 779 | 79 | |
| XM_009294035.3 | XP_009292310.1 | 7 - 8 | 1285 | 79 | ||
| XM_009294037.3 | XP_009292312.1 | 6 - 7 | 693 | 79 | ||
| GenBank mRNA | BC065869.1 | - | 8 - 9 | 779 | 79 | |
More Information
Set Membership: |
|

u2af2b
Genomic Map