Assay Details
| Gene Symbol: traf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205762.1 | NP_991325.1 | 4 - 5 | 718 | 88 | |
| GenBank mRNA | AJ556548.1 | - | 4 - 5 | 781 | 88 | |
| BC065969.1 | - | 4 - 5 | 718 | 88 | ||
More Information
Set Membership: |
|

traf4a
Genomic Map