Assay Details
| Gene Symbol: mbd3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212580.1 | NP_997745.1 | 3 - 4 | 402 | 85 | |
| XM_005170432.4 | XP_005170489.1 | 5 - 6 | 652 | 85 | ||
| XM_005170433.4 | XP_005170490.1 | 5 - 6 | 649 | 85 | ||
| GenBank mRNA | AY238338.1 | - | 3 - 4 | 402 | 85 | |
| BC093115.1 | - | 4 - 5 | 434 | 85 | ||
More Information
Set Membership: |
|

mbd3b
Genomic Map