Assay Details
| Gene Symbol: rnf128a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212615.1 | NP_997780.1 | 1 - 2 | 532 | 75 | |
| GenBank mRNA | BC067341.1 | - | 1 - 2 | 532 | 75 | |
More Information
Set Membership: |
|

rnf128a
Genomic Map