Assay Details
| Gene Symbol: orc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212671.2 | NP_997836.1 | 4 - 5 | 486 | 84 | |
| GenBank mRNA | BC056528.1 | - | 4 - 5 | 486 | 84 | |
| BC065625.1 | - | 4 - 5 | 469 | 84 | ||
More Information
Set Membership: |
|

orc6
Genomic Map