Assay Details
| Gene Symbol: rhot1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212704.1 | NP_997869.1 | 18 - 19 | 2050 | 84 | |
| GenBank mRNA | BC044431.1 | - | 18 - 19 | 2045 | 84 | |
| BC068190.1 | - | 18 - 19 | 2050 | 84 | ||
More Information
Set Membership: |
|

rhot1a
Genomic Map