Assay Details
| Gene Symbol: rhot1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212704.1 | NP_997869.1 | 9 - 10 | 945 | 115 | |
| XM_009299425.3 | XP_009297700.1 | 9 - 10 | 974 | 115 | ||
| XM_009299426.3 | XP_009297701.1 | 9 - 10 | 973 | 115 | ||
| XM_009299427.3 | XP_009297702.1 | 9 - 10 | 974 | 115 | ||
| GenBank mRNA | BC068190.1 | - | 9 - 10 | 945 | 115 | |
More Information
Set Membership: |
|

rhot1a
Genomic Map